4-(3-Bromo-4-chlorophenyl)-1-methyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

575520-5G 575520-25G 575520-1G 575520-500MG
77630-456EA 1484.23 USD
77630-456 77636-162 77639-738 77647-748
4-(3-Bromo-4-chlorophenyl)-1-methyl-1H-pyrazole ≥97%
4-(3-Bromo-4-chlorophenyl)-1-methyl-1H-pyrazole

Precaución: For Research Use Only

Formula: 271.5409 g/mol
MW: C₁₀H₈BrClN₂
Storage Temperature: Ambient
CAS Number: 1407884-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR