4-(5-Bromo-2-fluorophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

575500-1G 575500-500MG 575500-5G
77635-216EA 998 USD
77635-216 77653-040 77656-616
4-(5-Bromo-2-fluorophenyl)pyridine ≥97%
4-(5-Bromo-2-fluorophenyl)pyridine

Precaución: For Research Use Only

Formula: C₁₁H₇BrFN
Storage Temperature: Ambient
CAS Number: 425379-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR