4-(3-Bromo-5-(tert-butyl)phenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

575486-5G 575486-1G 575486-500MG
77660-124EA 2568 USD
77660-124 77643-802 77651-734
4-(3-Bromo-5-(tert-butyl)phenyl)pyridine ≥97%
4-(3-Bromo-5-(tert-butyl)phenyl)pyridine

Precaución: For Research Use Only

Formula: C₁₅H₁₆BrN
MW: 290.1982 g/mol
Storage Temperature: Ambient
CAS Number: 2990384-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR