1-(4-Chloro-2,3-difluorophenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

57347-5G 57347-1G 57347-25G 57347-500MG
77630-446EA 1484.23 USD
77630-446 77639-732 77656-608 77651-732
1-(4-Chloro-2,3-difluorophenyl)cyclopentanol ≥95%
1-(4-Chloro-2,3-difluorophenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 232.654 g/mol
MW: C₁₁H₁₁ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD34758149
CAS Number: 2918878-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR