Ethyl 3-(3-chlorophenyl)-3-hydroxypropanoate ≥95%

Supplier: AOBChem USA

Warning

57277-5G 57277-25G 57277-1G
77635-214EA 248 USD
77635-214 77656-606 77643-798
Ethyl 3-(3-chlorophenyl)-3-hydroxypropanoate ≥95%
Ethyl 3-(3-chlorophenyl)-3-hydroxypropanoate

Precaución: For Research Use Only

Formula: 228.672 g/mol
MW: C₁₁H₁₃ClO₃
Storage Temperature: Ambient
MDL Number: MFCD16749033
CAS Number: 51699-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR