1-(4-Fluorophenoxy)-3-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

572176-1G 572176-5G 572176-500MG
77639-720EA 694 USD
77639-720 77656-598 77651-722
1-(4-Fluorophenoxy)-3-nitrobenzene ≥97%
1-(4-Fluorophenoxy)-3-nitrobenzene

Precaución: For Research Use Only

Formula: C₁₂H₈FNO₃
Storage Temperature: Ambient
CAS Number: 263349-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR