1,5-Dimethyl-1H-pyrazole-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

57066-5G 57066-25G
77635-212EA 65 USD
77635-212 77660-112
1,5-Dimethyl-1H-pyrazole-4-carboxylic acid ≥97%
1,5-Dimethyl-1H-pyrazole-4-carboxylic acid

Precaución: For Research Use Only

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02090876
CAS Number: 31728-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR