2-Bromo-6-(tert-pentyl)phenol ≥97%

Supplier: AOBChem USA

Warning

57060-5G 57060-500MG 57060-25G 57060-1G
77630-428EA 1484.23 USD
77630-428 77653-036 77660-110 77643-788
2-Bromo-6-(tert-pentyl)phenol ≥97%
2-Bromo-6-(tert-pentyl)phenol

Precaución: For Research Use Only

Formula: 243.14 g/mol
MW: C₁₁H₁₅BrO
Storage Temperature: Ambient
MDL Number: MFCD35164940
CAS Number: 1334501-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR