4-Butyl-1,2-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

568531-5G 568531-25G 568531-1G 568531-500MG
77630-426EA 1484.23 USD
77630-426 77636-152 77643-786 77647-720
4-Butyl-1,2-dimethylbenzene ≥97%
4-Butyl-1,2-dimethylbenzene

Precaución: For Research Use Only

Formula: 162.2713 g/mol
MW: C₁₂H₁₈
Storage Temperature: Ambient
CAS Number: 15880-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR