1-(5-Chloro-3-fluoro-2-methoxyphenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
56841-5G
56841-25G
56841-1G
56841-500MG
77633-600EA
1059
USD
77633-600
77660-102
77643-776
77647-710
1-(5-Chloro-3-fluoro-2-methoxyphenyl)ethanone ≥97%
1-(5-Chloro-3-fluoro-2-methoxyphenyl)ethanone
Precaución: For Research Use Only
Formula:
202.61 g/mol MW: C₉H₈ClFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD28526630 CAS Number: 1781675-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...