1-(Benzyloxy)-2,4-difluoro-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

56762-5G 56762-25G 56762-1G 56762-500MG
77633-596EA 698 USD
77633-596 77656-578 77643-768 77651-702
1-(Benzyloxy)-2,4-difluoro-3-methylbenzene ≥95%
1-(Benzyloxy)-2,4-difluoro-3-methylbenzene

Precaución: For Research Use Only

Formula: C₁₄H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD34757930
CAS Number: 2918866-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR