2-(2-Fluoro-3-(methylthio)-5-(trifluoromethyl)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

56679-5G 56679-25G 56679-1G 56679-500MG
77630-408EA 1484.23 USD
77630-408 77656-574 77643-766 77647-704
2-(2-Fluoro-3-(methylthio)-5-(trifluoromethyl)phenyl)propan-2-ol ≥95%
2-(2-Fluoro-3-(methylthio)-5-(trifluoromethyl)phenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₂F₄OS
Storage Temperature: Ambient
MDL Number: MFCD34757883
CAS Number: 2918936-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR