1-(2-Fluoro-3-(methylthio)-5-(trifluoromethyl)phenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

56678-5G 56678-1G 56678-25G 56678-500MG
77630-406EA 1484.23 USD
77630-406 77639-696 77660-098 77647-702
1-(2-Fluoro-3-(methylthio)-5-(trifluoromethyl)phenyl)ethanol ≥95%
1-(2-Fluoro-3-(methylthio)-5-(trifluoromethyl)phenyl)ethanol

Precaución: For Research Use Only

Formula: 254.244 g/mol
MW: C₁₀H₁₀F₄OS
Storage Temperature: Ambient
MDL Number: MFCD34757882
CAS Number: 2918858-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR