5-Bromo-2-fluoro-4-iodophenol ≥97%

Supplier: AOBChem USA

Warning

56607-5G 56607-25G 56607-1G 56607-500MG
77633-594EA 733 USD
77633-594 77656-572 77647-700 77651-700
5-Bromo-2-fluoro-4-iodophenol ≥97%
5-Bromo-2-fluoro-4-iodophenol

Precaución: For Research Use Only

Formula: 316.894 g/mol
MW: C₆H₃BrFIO
Storage Temperature: Ambient
MDL Number: MFCD04038230
CAS Number: 530141-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR