2-Bromo-4-fluoro-1,3-diiodobenzene ≥97%

Supplier: AOBChem USA

Warning

56596-5G 56596-25G
77635-206EA 98 USD
77635-206 77656-570
2-Bromo-4-fluoro-1,3-diiodobenzene ≥97%
2-Bromo-4-fluoro-1,3-diiodobenzene

Precaución: For Research Use Only

Formula: 426.791 g/mol
MW: C₆H₂BrFI₂
Storage Temperature: Ambient
MDL Number: MFCD28738395
CAS Number: 1805105-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR