Methyl 4-bromo-2-formylbenzoate ≥95%

Supplier: AOBChem USA

Warning

56478-5G 56478-1G 56478-25G 56478-500MG
77635-204EA 395 USD
77635-204 77639-694 77660-096 77651-698
Methyl 4-bromo-2-formylbenzoate ≥95%
Methyl 4-bromo-2-formylbenzoate

Precaución: For Research Use Only

Formula: C₉H₇BrO₃
MW: 243.054 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16036574
CAS Number: 1260795-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR