5-Chloro-1-(2-methoxyphenyl)pentan-1-one ≥95%

Supplier: AOBChem USA

Warning

564489-5G 564489-1G 564489-500MG 564489-25G
77633-592EA 1438 USD
77633-592 77639-692 77653-034 77656-568
5-Chloro-1-(2-methoxyphenyl)pentan-1-one ≥95%
5-Chloro-1-(2-methoxyphenyl)pentan-1-one

Precaución: For Research Use Only

Formula: C₁₂H₁₅ClO₂
Storage Temperature: Ambient
CAS Number: 43228-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR