5-Chloro-2-fluoro-3-methylphenol ≥97%

Supplier: AOBChem USA

Warning

56401-5G 56401-1G 56401-25G 56401-500MG
77633-586EA 987 USD
77633-586 77639-682 77656-558 77647-688
5-Chloro-2-fluoro-3-methylphenol ≥97%
5-Chloro-2-fluoro-3-methylphenol

Precaución: For Research Use Only

Formula: C₇H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD28516577
CAS Number: 1781655-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR