Methyl 4-hydroxy-2,6-dimethylbenzoate ≥95%

Supplier: AOBChem USA

Warning

56324-5G 56324-25G
77635-200EA 173 USD
77635-200 77656-556
Methyl 4-hydroxy-2,6-dimethylbenzoate ≥95%
Methyl 4-hydroxy-2,6-dimethylbenzoate

Precaución: For Research Use Only

Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD13191754
CAS Number: 83194-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR