1-(4-Fluoro-2,5-dimethylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

562669-5G 562669-1G 562669-500MG
77630-394EA 1484.23 USD
77630-394 77639-676 77647-684
1-(4-Fluoro-2,5-dimethylphenyl)ethanone ≥95%
1-(4-Fluoro-2,5-dimethylphenyl)ethanone

Precaución: For Research Use Only

Formula: 166.192 g/mol
MW: C₁₀H₁₁FO
Storage Temperature: Ambient
CAS Number: 42444-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR