4-Bromo-1,2,3,6-tetrahydro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

562487-5G 562487-25G 562487-1G 562487-500MG
77630-392EA 1484.23 USD
77630-392 77636-144 77643-754 77647-682
4-Bromo-1,2,3,6-tetrahydro-1,1'-biphenyl ≥97%
4-Bromo-1,2,3,6-tetrahydro-1,1'-biphenyl

Precaución: For Research Use Only

Formula: 237.1356 g/mol
MW: C₁₂H₁₃Br
Storage Temperature: Ambient
CAS Number: 1247868-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR