1-Bromo-4-pentylcyclohexene ≥97%

Supplier: AOBChem USA

Warning

562484-5G 562484-25G
77635-196EA 171 USD
77635-196 77660-082
1-Bromo-4-pentylcyclohexene ≥97%
1-Bromo-4-pentylcyclohexene

Precaución: For Research Use Only

Formula: C₁₁H₁₉Br
Storage Temperature: Ambient
MDL Number: MFCD10699694
CAS Number: 1033202-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR