2-Chloro-4-morpholinobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

558561-5G 558561-1G 558561-25G 558561-500MG
77635-194EA 601 USD
77635-194 77639-672 77656-552 77651-686
2-Chloro-4-morpholinobenzonitrile ≥97%
2-Chloro-4-morpholinobenzonitrile

Precaución: For Research Use Only

Formula: C₁₁H₁₁ClN₂O
Storage Temperature: Ambient
CAS Number: 20925-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR