2-(4-Bromo-3-fluorophenyl)acetamide ≥95%

Supplier: AOBChem USA

Warning

556748-5G 556748-25G 556748-1G 556748-500MG
77630-382EA 1484.23 USD
77630-382 77656-546 77643-742 77651-682
2-(4-Bromo-3-fluorophenyl)acetamide ≥95%
2-(4-Bromo-3-fluorophenyl)acetamide

Precaución: For Research Use Only

Formula: 232.0496 g/mol
MW: C₈H₇BrFNO
Storage Temperature: Ambient
CAS Number: 1851769-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR