4-Ethynyl-2-fluoro-5-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

556040-5G 556040-25G 556040-1G 556040-500MG
77630-380EA 1484.23 USD
77630-380 77636-138 77639-664 77647-668
4-Ethynyl-2-fluoro-5-methoxypyridine ≥97%
4-Ethynyl-2-fluoro-5-methoxypyridine

Precaución: For Research Use Only

Formula: 151.1377 g/mol
MW: C₈H₆FNO
Storage Temperature: Ambient
CAS Number: 1824076-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR