2-Fluoro-4-((trimethylsilyl)ethynyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

556023-5G 556023-500MG 556023-25G 556023-1G
77630-378EA 1484.23 USD
77630-378 77653-028 77660-068 77643-738
2-Fluoro-4-((trimethylsilyl)ethynyl)pyridine ≥95%
2-Fluoro-4-((trimethylsilyl)ethynyl)pyridine

Precaución: For Research Use Only

Formula: 193.2928 g/mol
MW: C₁₀H₁₂FNSi
Storage Temperature: Ambient
CAS Number: 1378976-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR