3-(5-Isopropyl-2-methoxyphenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

555858-5G 555858-1G 555858-500MG
77660-064EA 2245 USD
77660-064 77643-736 77651-676
3-(5-Isopropyl-2-methoxyphenyl)oxetan-3-ol ≥95%
3-(5-Isopropyl-2-methoxyphenyl)oxetan-3-ol

Precaución: For Research Use Only

Formula: 222.28 g/mol
MW: C₁₃H₁₈O₃
Storage Temperature: Ambient
CAS Number: 1903450-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR