1-(3-Ethyl-2-hydroxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

555613-5G 555613-500MG 555613-25G 555613-1G
77633-576EA 886 USD
77633-576 77653-026 77656-540 77643-732
1-(3-Ethyl-2-hydroxyphenyl)ethanone ≥97%
1-(3-Ethyl-2-hydroxyphenyl)ethanone

Precaución: For Research Use Only

Formula: 164.2010 g/mol
MW: C₁₀H₁₂O₂
Storage Temperature: Ambient
CAS Number: 103323-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR