(6-Chloro-2-fluoropyridin-3-yl)(cyclopropyl)methanone ≥95%

Supplier: AOBChem USA

Warning

554995-5G 554995-25G 554995-1G 554995-500MG
77630-340EA 1736.58 USD
77630-340 77656-530 77643-718 77651-654
(6-Chloro-2-fluoropyridin-3-yl)(cyclopropyl)methanone ≥95%
(6-Chloro-2-fluoropyridin-3-yl)(cyclopropyl)methanone

Precaución: For Research Use Only

Formula: C₉H₇ClFNO
Storage Temperature: Ambient
CAS Number: 1662713-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR