1-(6-Chloro-2-fluoropyridin-3-yl)cyclobutanol ≥97%

Supplier: AOBChem USA

Warning

554992-5G 554992-1G 554992-25G 554992-500MG
77630-336EA 1725.25 USD
77630-336 77639-640 77656-526 77651-650
1-(6-Chloro-2-fluoropyridin-3-yl)cyclobutanol ≥97%
1-(6-Chloro-2-fluoropyridin-3-yl)cyclobutanol

Precaución: For Research Use Only

Formula: C₉H₉ClFNO
Storage Temperature: Ambient
CAS Number: 1831086-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR