3,6-Dichloro-2-fluoropyridine ≥95%

Supplier: AOBChem USA

Warning

554990-5G 554990-1G 554990-25G 554990-500MG
77633-570EA 879 USD
77633-570 77639-638 77660-046 77647-646
3,6-Dichloro-2-fluoropyridine ≥95%
3,6-Dichloro-2-fluoropyridine

Precaución: For Research Use Only

Formula: C₅H₂Cl₂FN
Storage Temperature: Ambient
CAS Number: 51991-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR