Benzo[b]thiophen-2-yl(cyclopropyl)methanone ≥95%

Supplier: AOBChem USA

Warning

554981-5G 554981-1G 554981-500MG
77656-518EA 2073 USD
77656-518 77647-640 77651-642
Benzo[b]thiophen-2-yl(cyclopropyl)methanone ≥95%
Benzo[b]thiophen-2-yl(cyclopropyl)methanone

Precaución: For Research Use Only

Formula: C₁₂H₁₀OS
Storage Temperature: Ambient
CAS Number: 1603561-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR