6-Phenoxynicotinaldehyde oxime ≥95%

Supplier: AOBChem USA

Warning

554324-5G 554324-25G 554324-1G 554324-500MG
77633-562EA 1201 USD
77633-562 77656-512 77643-696 77647-634
6-Phenoxynicotinaldehyde oxime ≥95%
6-Phenoxynicotinaldehyde oxime

Precaución: For Research Use Only

Formula: 214.22 g/mol
MW: C₁₂H₁₀N₂O₂
Storage Temperature: Ambient
CAS Number: 1355245-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR