4,5-Dibromo-2-methylaniline ≥97%

Supplier: AOBChem USA

Warning

55400-5G 55400-500MG 55400-25G 55400-1G
77635-192EA 438 USD
77635-192 77653-020 77660-034 77643-694
4,5-Dibromo-2-methylaniline ≥97%
4,5-Dibromo-2-methylaniline

Precaución: For Research Use Only

Formula: C₇H₇Br₂N
Storage Temperature: Ambient
MDL Number: MFCD11845874
CAS Number: 102170-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR