1-(4-Fluorophenoxy)-2-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

552617-1G 552617-500MG 552617-5G
77635-188EA 975 USD
77635-188 77653-018 77660-032
1-(4-Fluorophenoxy)-2-nitrobenzene ≥97%
1-(4-Fluorophenoxy)-2-nitrobenzene

Precaución: For Research Use Only

Formula: C₁₂H₈FNO₃
Storage Temperature: Ambient
CAS Number: 4475-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR