4'-Bromo-2',5'-dimethyl-[1,1'-biphenyl]-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

55037-5G 55037-1G 55037-500MG
77630-314EA 1738.64 USD
77630-314 77643-690 77647-626
4'-Bromo-2',5'-dimethyl-[1,1'-biphenyl]-3-carbaldehyde ≥97%
4'-Bromo-2',5'-dimethyl-[1,1'-biphenyl]-3-carbaldehyde

Precaución: For Research Use Only

Formula: 289.167 g/mol
MW: C₁₅H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD34530349
CAS Number: 2734776-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR