2,2-Dichloro-1-(3-chlorophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

54989-5G 54989-1G 54989-25G 54989-500MG
77635-186EA 488 USD
77635-186 77639-624 77656-506 77651-626
2,2-Dichloro-1-(3-chlorophenyl)ethanol ≥95%
2,2-Dichloro-1-(3-chlorophenyl)ethanol

Precaución: For Research Use Only

Formula: C₈H₇Cl₃O
Storage Temperature: Ambient
MDL Number: MFCD00018865
CAS Number: 27683-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR