1-(4-Bromo-2-ethylphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

549645-5G 549645-1G 549645-25G 549645-500MG
77633-560EA 1357 USD
77633-560 77639-622 77660-028 77651-624
1-(4-Bromo-2-ethylphenyl)ethanol ≥97%
1-(4-Bromo-2-ethylphenyl)ethanol

Precaución: For Research Use Only

Formula: 229.1136 g/mol
MW: C₁₀H₁₃BrO
Storage Temperature: Ambient
CAS Number: 1892277-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR