2-(Methylthio)pyrimidine-5-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

54951-5G 54951-1G 54951-25G 54951-500MG
77635-182EA 316 USD
77635-182 77639-620 77660-026 77651-622
2-(Methylthio)pyrimidine-5-carboxylic acid ≥97%
2-(Methylthio)pyrimidine-5-carboxylic acid

Precaución: For Research Use Only

Formula: C₆H₆N₂O₂S
MW: 170.1890 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07394096
CAS Number: 110099-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR