N-(4-chlorophenyl)benzimidamide ≥95%

Supplier: AOBChem USA

Warning

549376-5G 549376-25G 549376-1G 549376-500MG
77630-308EA 1642.85 USD
77630-308 77636-114 77643-682 77647-622
N-(4-chlorophenyl)benzimidamide ≥95%
N-(4-chlorophenyl)benzimidamide

Precaución: For Research Use Only

Formula: C₁₃H₁₁ClN₂
Storage Temperature: Ambient
CAS Number: 7035-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR