4'-Fluoro-3',5'-dimethylacetophenone ≥97%

Supplier: AOBChem USA

Warning

54872-5G 54872-25G 54872-1G
77635-180EA 234 USD
77635-180 77660-024 77643-680
4'-Fluoro-3',5'-dimethylacetophenone ≥97%
4'-Fluoro-3',5'-dimethylacetophenone

Precaución: For Research Use Only

Formula: C₁₀H₁₁FO
MW: 166.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660269
CAS Number: 42444-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR