2,3,6-Trichlorophenol ≥97%

Supplier: AOBChem USA

Warning

54788-5G 54788-25G
77635-178EA 86 USD
77635-178 77656-500
2,3,6-Trichlorophenol ≥97%
2,3,6-Trichlorophenol

Precaución: For Research Use Only

Formula: C₆H₃Cl₃O
Storage Temperature: Ambient
MDL Number: MFCD00002165
CAS Number: 933-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR