1,3-Dichloro-2-methyl-4-phenoxybenzene ≥95%

Supplier: AOBChem USA

Warning

546814-5G 546814-1G 546814-25G 546814-500MG
77630-288EA 1818.98 USD
77630-288 77639-608 77660-016 77651-606
1,3-Dichloro-2-methyl-4-phenoxybenzene ≥95%
1,3-Dichloro-2-methyl-4-phenoxybenzene

Precaución: For Research Use Only

Formula: C₁₃H₁₀Cl₂O
Storage Temperature: Ambient
CAS Number: 68486-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR