2-(4-Fluoro-2-methoxy-5-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

546164-5G 546164-1G 546164-500MG
77656-490EA 2083 USD
77656-490 77643-662 77651-600
2-(4-Fluoro-2-methoxy-5-methylphenyl)propan-2-ol ≥95%
2-(4-Fluoro-2-methoxy-5-methylphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 198.234 g/mol
MW: C₁₁H₁₅FO₂
Storage Temperature: Ambient
CAS Number: 1891401-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR