1-((2-Bromophenoxy)methyl)-2-chloro-4-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

545430-1G 545430-5G 545430-500MG
77639-600EA 560 USD
77639-600 77660-010 77647-600
1-((2-Bromophenoxy)methyl)-2-chloro-4-fluorobenzene ≥97%
1-((2-Bromophenoxy)methyl)-2-chloro-4-fluorobenzene

Precaución: For Research Use Only

Formula: C₁₃H₉BrClFO
Storage Temperature: Ambient
CAS Number: 1041518-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR