4-Chlorodibenzo[b,d]furan ≥97%

Supplier: AOBChem USA

Warning

54528-5G 54528-25G 54528-1G 54528-500MG
77633-554EA 1284 USD
77633-554 77656-488 77643-660 77651-598
4-Chlorodibenzo[b,d]furan ≥97%
4-Chlorodibenzo[b,d]furan

Precaución: For Research Use Only

Formula: C₁₂H₇ClO
Storage Temperature: Ambient
MDL Number: MFCD00800122
CAS Number: 74992-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR