4-Chloro-1-(m-tolyl)butan-1-one ≥95%

Supplier: AOBChem USA

Warning

545084-5G 545084-1G 545084-25G 545084-500MG
77633-550EA 1357 USD
77633-550 77639-598 77656-486 77647-596
4-Chloro-1-(m-tolyl)butan-1-one ≥95%
4-Chloro-1-(m-tolyl)butan-1-one

Precaución: For Research Use Only

Formula: 196.673 g/mol
MW: C₁₁H₁₃ClO
Storage Temperature: Ambient
CAS Number: 40877-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR