3-Methyl-1-(m-tolyl)but-2-en-1-one ≥95%

Supplier: AOBChem USA

Warning

545083-5G 545083-1G 545083-500MG
77656-484EA 2001 USD
77656-484 77643-656 77647-594
3-Methyl-1-(m-tolyl)but-2-en-1-one ≥95%
3-Methyl-1-(m-tolyl)but-2-en-1-one

Precaución: For Research Use Only

Formula: 174.239 g/mol
MW: C₁₂H₁₄O
Storage Temperature: Ambient
CAS Number: 1253905-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR