4-Chloro-3-cyano-2-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

545064-5G 545064-25G 545064-1G 545064-500MG
77633-548EA 969 USD
77633-548 77660-004 77643-650 77651-588
4-Chloro-3-cyano-2-fluorobenzoic acid ≥95%
4-Chloro-3-cyano-2-fluorobenzoic acid

Precaución: For Research Use Only

Formula: C₈H₃ClFNO₂
Storage Temperature: Ambient
CAS Number: 1628684-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR