(2,3-Dichloro-4-(trifluoromethyl)phenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

544720-5G 544720-25G 544720-1G 544720-500MG
77633-544EA 1441 USD
77633-544 77656-476 77643-644 77647-586
(2,3-Dichloro-4-(trifluoromethyl)phenyl)boronic acid ≥95%
(2,3-Dichloro-4-(trifluoromethyl)phenyl)boronic acid

Precaución: For Research Use Only

Formula: 258.818 g/mol
MW: C₇H₄BCl₂F₃O₂
Storage Temperature: Ambient
CAS Number: 2940986-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR